Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamate Receptor Mouse, Unlabeled, Clone: 6C4, BD

Catalog No. BDB556341 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556341 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556341 Supplier BD Biosciences Supplier No. 556341

Mouse Monoclonal Antibody

Glutamate is a major excitatory neurotransmitter in mammalian brain. Glutametergic neurotransmission is mediated by a family of glutamate receptors which can be grouped in two classes, ionotropic (GluR) and metabotropic (mGluR) receptors. The ionotropic GluRs can be divided into two subclasses, N-methyl-D-Aspartate (NMDA) and non-NMDA receptors. Five different forms of NMDA receptors (NMDAR1, R2A, R2B, R2C, and R2D) have been isolated. NMDAR1 is always required for the formation of functional NMDA receptors. Non-NMDA receptors can be divided into at least two subtypes, AMPA receptors which bind to methyl-4-isoxazole proprionic acid (AMPA) and kainate binding (KA or kainic acid) receptors. Multiple subunits appear to comprise the family of non-NMDA receptors. GluR1-4 receptors (also known as GluR-A, B, C, and D) preferentially bind to AMPA. KA receptors consist of five subunits: GluR5, 6, 7, KA-1, and KA-2. Functional receptors are formed by various combinations of these subunits and multiple forms of GluR are expressed in the same populations
of neurons. GluR2 migrates at a molecular weight of ∽102kDa in SDS-PAGE. Monoclonal antibody (mAb) 6C4 recognizes rat, macaque monkey, and dog GluR2. It does not cross-react with GluRs 1, 3, 4c, 5, 6, or 7. A recombinant trpE fusion protein containing the putative N-terminal portion (amino acids 175-430) of GluR2 was used as immunogen. The antibody was originally characterized by western blot analysis and immunohistochemical analysis of paraformaldehyde-fixed cells and paraformaldehyde-fixed frozen tissue sections. Its specificity for GluR2 was determined by radioimmunoassay and by western blot analysis. In western blots obtained from cells transiently transfected from with different GluRs, the antibody recognized GluR2 but not GluRs 1, 3, 4c, 5, 6, 7. Cross-species western blot reactivity was determined by using homogenates of fresh frozen tissue samples from rat, macaque monkey and dog cerebral cortex. In western blot analysis of tissue homogenates, clone 6C4 recognizes GluR2 as an ∽102kDa protein. Other proteins noted by western blot at 66kDa and lower molecular weights appear to be breakdown products of GluR2. The specificity of 6C4 for GluR2 in rat brain tissue sections was verified by showing that antibody reactivity was blocked by preincubation with recombinant GluR2 protein but not with recombinant GluR1 or GluR3 protein. Immunohistochemical analysis using 6C4 shows that GluR2 is widely distributed at both the cellular and synaptic levels in rat hippocampus and neocortex, please refer to Vissavajjhala, et al. for additional staining information.

Host Species: Mouse
Clone: 6C4
Isotype: IgG2a κ
Species Reactivity: Rat
Immunogen: GluR2 aa.175-430 trpE Recombinant Fusion

Immunohistochemistry, Radioimmunoassay, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamate Receptor
Applications Immunohistochemistry
Classification Monoclonal
Clone 6C4
Concentration 0.5mg/mL
Conjugate Unconjugated
Description GluR2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen GluR2 aa.175-430 trpE Recombinant Fusion
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Monkey, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.