Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamate Receptor Delta 1/2 Rabbit anti-Human, Mouse, Primate, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB2285MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2285MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2285MI Supplier MilliporeSigma Supplier No. AB2285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Glutamate Receptor Delta 1/2 Antibody detects level of Glutamate Receptor Delta 1/2 and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamate Receptor Delta 1/2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.
Gene Symbols GRID1; KIAA1220 ; OTTHUMP00000059648
Host Species Rabbit
Immunogen Synthetic linear peptide to the C-Terminus, conjugated to BSA.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_017551.2
Target Species Human, Mouse, Primate, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.