Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Glutamate Transporter 1 (GLT-1) Ab-1, Polyclonal, Invitrogen™
Supplier: Thermo Scientific PA3040
Description
Host Species: Rabbit
Cross Reactivity: Rt
Immunoprecipitation, Western Blotting

Specifications
Glutamate Transporter 1 (GLT-1) | |
Unconjugated | |
100 μL | |
Rat |
Polyclonal | |
Rabbit | |
Primary |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction