Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamate Transporter Mouse anti-Rat, Clone: 4D6.2, MilliporeSigma™

Catalog No. MAB1587MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1587MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1587MI Supplier MilliporeSigma Supplier No. MAB1587
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Glutamate Transporter Antibody, neuronal, clone 4D6.2 is an antibody against Glutamate Transporter for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamate Transporter
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4D6.2
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M Phosphate buffer, 0.25M NaCl, pH 7.6 with 0.1% sodium azide.
Gene Accession No. NM_004170.4
Gene Symbols SLC1A1, EAAC1, EAAT3
Host Species Mouse
Immunogen C-terminus peptide (KDKSDTISFTQTSQF) (Catalog Number AG372).
Quantity 100 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage 2-8°C for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.