Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamate Transporter Guinea Pig anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1783MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1783MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1783MI Supplier MilliporeSigma Supplier No. AB1783
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Detect Glutamate Transporter using this Anti-Glutamate Transporter Antibody, Glial validated for use in immunohistochemistry (paraffin), immunofluorescence and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamate Transporter, Glial
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Guinea pig antiserum containing no preservatives.
Gene Symbols SLC1A2; GLT-1; EAAT2; GLT1
Host Species Guinea Pig
Immunogen Synthetic peptide from the carboxy-terminus of rat GLT-1.
Purification Method Unpurified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_004171.3
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.