Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamine Synthetase Mouse, Unlabeled, Clone: 6, BD

Catalog No. BDB610518 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610518 150 μg
BDB610517 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610518 Supplier BD Biosciences Supplier No. 610518
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glutamine synthetase catalyzes the amination of glutamic acid to form glutamine. It is found in mammals as an octamer of identical 45 kDa subunits. Glutamine synthetase activity is a useful marker for astrocytes and an important differentiation feature in retina. It is also considered to be a key enzyme in the recycling of the neurotransmitter glutamate.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamine Synthetase
Applications Western Blot
Classification Monoclonal
Clone 6
Concentration 250μg/mL
Conjugate Unconjugated
Description Glutamate-Ammonia Ligase; GLUL; GLNS
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.