Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamine Synthetase Mouse anti-Human, Mouse, Rat, Sheep, Clone: GS-6, MilliporeSigma™

Catalog No. MAB302MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB302MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB302MI Supplier MilliporeSigma Supplier No. MAB302
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Glutamine Synthetase using this Anti-Glutamine Synthetase Antibody, clone GS-6 validated for use in ELISA, immunohistochemistry, immunohistochemistry (paraffin) and western blotting with more than 45 product citations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamine Synthetase
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone GS-6
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in buffer containing 50% glycerol, BSA and 0.09% sodium azide.
Gene Accession No. NM_001033044.1
Gene Symbols GLUL, GLNS
Host Species Mouse
Immunogen Glutamine synthetase purified from sheep brain.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Sheep
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.