Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione: N-ethylmaleimide adduct Mouse anti-Mammals, Clone: 8.1GSH, MilliporeSigma™

Catalog No. MAB3194MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3194MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3194MI Supplier MilliporeSigma Supplier No. MAB3194
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect the Glutathione: N-ethylmaleimide adduct protein using this Anti-Glutathione: N-ethylmaleimide adduct, clone 8.1GSH validated for use in ELISA, Flow, immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutathione: N-ethylmaleimide adduct
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8.1GSH
Conjugate Unconjugated
Formulation Liquid in 0.02M PBS pH 7.6, 0.25M NaCl containing 0.1% sodium azide.
Host Species Mouse
Immunogen GS-NEM conjugated to KLH
Quantity 100 μg
Primary or Secondary Primary
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.