Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione-S-Transferase Rabbit anti-Vertebrates, Schistosoma japonicum, Polyclonal, MilliporeSigma™

Catalog No. AB3282MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3282MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3282MI Supplier MilliporeSigma Supplier No. AB3282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Glutathione-S-Transferase Antibody, S. japonicum form is an antibody against Glutathione-S-Transferase for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutathione-S-Transferase
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in PBS, pH 7.4, containing 0.02% Tween 20 and 0.02% sodium azide as a preservative.
Gene Symbols GSTM1; MGC26563; GSTM1-1; H-B; GSTM1b-1b; GTH4; GSTM1a-1a; GTM1; MU; MU-1; GST1; EC 2.5.1.18
Host Species Rabbit
Immunogen Recombinant glutathione S-transferase (GST) protein from Schistosoma japonicum.
Quantity 500 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000561.2; NM_146421.1
Target Species S. japonicum, Vertebrates
Content And Storage 2-8°C for up to 6 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.