Learn More
MilliporeSigma™ anti-Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH; G3PDH), Clone: 6C5
Supplier: MilliporeSigma™ MAB374/DEL
Description
Glyceraldehyde 3-phosphate dehydrogenase (GAPDH) is an ubiquitous glycolytic enzyme present in reasonably high levels in almost all tissues. As a 'house-keeping' enzyme, it catalyzes the synthesis of 1,3-biphosphoglycerate, a “high energy” intermediate used for the synthesis of ATP. Besides its cytoplasmic action in metabolism it is also involved in the initial stages of apoptosis or oxidative stress response where GAPDH is translocated to the nucleus. Such actions may reflect the role of GAPDH in DNA repair or as one nuclear carrier for apoptotic molecules. GAPDH has also been found to bind specifically to proteins implicated in the pathogenesis of a variety of neurodegenerative disorders including the beta-amyloid precursor protein and the huntingtin protein where decreased function of GAPDH is associated with Alzheimer's and Huntington's disease fibroblasts. GAPDH has also been identified as a potential target for nitric oxide (NO)-mediated cellular toxicity. The complete and functional enzyme is a tetramer with each of four identical subunits occupying the vertex of a tetrahedron. Binding domains also include one principally interacting with NAD+ and another interacting with glyceraldehyde 3-phosphate (GAP).
Affinity Purification, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blotting
Specifications
GAPDH (Glyceraldehyde-3-Phosphate Dehydrogenase) | |
Unconjugated | |
Glyceraldehyde-3-phosphate dehydrogenase from rabbit muscle | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
200 μg | |
Endocrine/Metabolism | |
Canine, Feline, Fish, Human, Mouse, Pig, Rabbit, Rat | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.