Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Glycerol kinase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527669
Description
Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (90%), Pig (97%), Chimpanzee (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The product of this gene belongs to the FGGY kinase family of proteins and encodes glycerol kinase. Glycerol kinase is a key enzyme in the regulation of glycerol uptake and metabolism. It catalyzes the phosphorylation of glycerol by ATP, yielding ADP and glycerol-3-phosphate. Defects in this gene are the cause of glycerol kinase deficiency (GKD). Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
Glycerol kinase | |
Polyclonal | |
Unconjugated | |
GK | |
ASTP; ATP:glycerol 3-phosphotransferase; ATP-stimulated glucocorticoid-receptor translocation promoter; ATP-stimulated glucocorticoid-receptor translocaton promoter; D930012N15Rik; EC 2.7.1.30; GK; GK 3; GK1; GK3P; GKD; GKl; GKP3; GKTB; glycerokinase; Glycerokinase 3; glycerol kinase; Glycerolkinase; Gyk | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2710 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.28 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P32189 | |
GK | |
Recombinant fragment corresponding to a region within amino acids 252 and 418 of Human Glycerol kinase. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction