Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine (Low Glutaraldehyde) Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5020MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5020MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5020MI Supplier MilliporeSigma Supplier No. AB5020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Glycine (Low Glutaraldehyde) using this Anti-Glycine (Low Glutaraldehyde) Antibody validated for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glycine (Low Glutaraldehyde)
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in stabilizing buffer (Phosphate buffer with sterile goat serum) containing 0.05% thimerosal.
Host Species Rabbit
Immunogen Glycine-glutaraldehyde-BSA
Quantity 500 μL
Primary or Secondary Primary
Target Species Rat
Content And Storage 2-8°C for up to 6 months, do not freeze
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.