Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB139MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB139MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB139MI Supplier MilliporeSigma Supplier No. AB139

Rabbit Polyclonal Antibody

This Anti-Glycine Antibody is validated for use in immunohistochemistry for the detection of Glycine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glycine
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit antiserum. Liquid with 0.05% sodium azide.
Host Species Rabbit
Immunogen Glycine-gluteraldehyde-BSA
Quantity 100 μL
Primary or Secondary Primary
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.