Learn More
Glycine Receptor α3 Subunit Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Glycine Receptor α3 Subunit |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 5% sucrose as a stabilizer and 0.05% sodium azide as a preservative. Reconstitute with 200μL of sterile deionized water. Centrifuge antibody prep |
| Gene Symbols | GLRA3 |
| Host Species | Rabbit |
| Immunogen | A purified peptide corresponding to amino acid residues 355-368 of rat GlyRa3 (Accession P24524). |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.