Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine Receptor α3 subunit Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB15014MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15014MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15014MI Supplier MilliporeSigma Supplier No. AB15014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Glycine Receptor α3 subunit Antibody detects level of Glycine Receptor α3 subunit and has been published and validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glycine Receptor α3 subunit
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% sodium azide.
Gene Symbols GLRA3
Host Species Rabbit
Immunogen This antibody is generated from rabbits immunized with a KLH conjugated linear peptide selected from residues surrounding the C-terminus of human Glycine Receptor α3 subunit.
Purification Method Affinity Purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_006520.2
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.