Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glypican-1 Mouse anti-Human, Rat, Clone: 4D1, MilliporeSigma™

Catalog No. MAB2600MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2600MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2600MI Supplier MilliporeSigma Supplier No. MAB2600
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Glypican-1 Antibody, clone 4D1 detects level of Glypican-1 and has been published and validated for use in immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glypican-1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4D1
Conjugate Unconjugated
Formulation Purified in 0.1MTris-Glycine (pH 7.4) 150mM NaCl with 0.05% NaN3.
Gene Accession No. NP_002072
Gene Symbols FLJ38078, glypican
Host Species Mouse
Immunogen Full length recombinant protein.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.