Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Rat anti-Human, Alexa Fluor 647, Clone: BVD2-21C11, BD

Catalog No. BDB562257 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562257 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562257 Supplier BD Biosciences Supplier No. 562257
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The BVD2-21C11 monoclonal antibody specifically binds to human Granulocyte/Macrophage - Colony Stimulating Factor (GM-CSF). Human GM-CSF is encoded by the CSF2 gene and is also known as Colony Stimulating Factor 2. GM-CSF is produced by activated T lymphocytes, macrophages, endothelial cells, fibroblasts, stromal cells and other cell types including B lymphocytes, mast cells, eosinophils, and osteoblasts. GM-CSF stimulates the survival, proliferation and/or differentiation of various cell types including neutrophils, eosinophils, macrophages, dendritic cells, megakaryocytes, erythroid cells, endothelial cells and their precursors. The immunogen used to generate the BVD2-21C11 hybridoma was recombinant human GM-CSF. The BVD2-21C11 antibody has been reported to crossreact with GM-CSF from the rhesus monkey. BVD2-21C11 is a neutralizing antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GM-CSF
Applications Flow Cytometry
Classification Monoclonal
Clone BVD2-21C11
Conjugate Alexa Fluor 647
Description CSF2; Colony stimulating factor 2 (granulocyte-macrophage); CSF; GMCSF
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant human GM-CSF
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.