Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Rat anti-Human, Unlabeled, Clone: BVD2-21C11, BD

Catalog No. BDB554503 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554503 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554503 Supplier BD Biosciences Supplier No. 554503
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The BVD2-21C11 antibody reacts with human granulocyte/macrophage - colony stimulating factor (GM-CSF). The immunogen used to generate the BVD2-21C11 hybridoma was recombinant human GM-CSF. The BVD2-21C11 antibody has been reported to cross react with GM-CSF from rhesus monkey. This is a neutralizing antibody.

Host Species: Rat
Clone: BVD2-21C11
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Recombinant human GM-CSF

Blocking Assay, Immunoprecipitation, Intracellular Staining, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GM-CSF
Applications Blocking Assay, Flow Cytometry
Classification Monoclonal
Clone BVD2-21C11
Concentration 0.5mg/mL
Conjugate Unconjugated
Description CSF2; Colony stimulating factor 2 (granulocyte-macrophage); CSF; GMCSF
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant human GM-CSF
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.