Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM-CSF Rat anti-Mouse, Unlabeled, Clone: MP1-22E9, BD

Catalog No. BDB554404 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554404 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554404 Supplier BD Biosciences Supplier No. 554404
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MP1-22E9 antibody reacts with mouse granulocyte/macrophage-colony stimulating factor (GM-CSF). The immunogen used to generate the MP1-22E9 hybridoma was yeast-expressed recombinant mouse GM-CSF. This is a neutralizing antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GM-CSF
Applications ELISA
Classification Monoclonal
Clone MP1-22E9
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant mouse GM-CSF
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.