Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GM2A, Mouse, Clone: 2C8, Abnova™

Catalog No. 89022338 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-338 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-338 Supplier Abnova Corporation Supplier No. H00002760M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GM2A.

This gene encodes a small glycolipid transport protein which acts as a substrate specific co-factor for the lysosomal enzyme beta-hexosaminidase A. Beta-hexosaminidase A, together with GM2 ganglioside activator, catalyzes the degradation of the ganglioside GM2, and other molecules containing terminal N-acetyl hexosamines. Mutations in this gene result in GM2-gangliosidosis type AB or the AB variant of Tay-Sachs disease. [provided by RefSeq

Sequence: WIKIPCTDYIGSCTFEHFCDVLDMLIPTGEPCPEPLRTYGLPCHCPFKEGTYSLPKSEFVVPDLELPSWLTTGNYRIESVLSSSGKRLGCIKIAASLKGI

Specifications

Antigen GM2A
Applications ELISA
Classification Monoclonal
Clone 2C8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GM2A.
Formulation PBS with no preservative; pH 7.4
Gene GM2A
Gene Accession No. NM_000405
Gene Alias SAP-3
Gene Symbols GM2A
Host Species Mouse
Immunogen GM2A (NP_000396.2, 94 a.a. ∼ 193 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2760
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.