Learn More
guanine monphosphate synthetase, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
In the de novo synthesis of purine nucleotides, IMP is the branch point metabolite at which point the pathway diverges to the synthesis of either guanine or adenine nucleotides. In the guanine nucleotide pathway, there are 2 enzymes involved in converting IMP to GMP, namely IMP dehydrogenase (IMPD1), which catalyzes the oxidation of IMP to XMP, and GMP synthetase, which catalyzes the amination of XMP to GMP. [provided by RefSeq
Specifications
Specifications
| Antigen | guanine monphosphate synthetase |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant GMPS. |
| Formulation | 50% glycerol |
| Gene | GMPS |
| Gene Accession No. | NM_003875 |
| Gene Symbols | GMPS |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.