Learn More
GNAQ, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human GNAQ peptide using ARM Technology.
Supplier: Abnova Corporation H00002776K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Guanine nucleotide-binding proteins are a family of heterotrimeric proteins that couple cell surface, 7-transmembrane domain receptors to intracellular signaling pathways. Receptor activation catalyzes the exchange of GTP for GDP bound to the inactive G protein alpha subunit resulting in a conformational change and dissociation of the complex. The G protein alpha and beta-gamma subunits are capable of regulating various cellular effectors. Activation is terminated by a GTPase intrinsic to the G-alpha subunit. G-alpha-q is the alpha subunit of one of the heterotrimeric GTP-binding proteins that mediates stimulation of phospholipase C-beta (MIM 600230).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
GNAQ | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
GNAQ | |
GNAQ | |
A synthetic peptide of human GNAQ | |
RUO | |
2776 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
guanine nucleotide binding protein (G protein), q polypeptide | |
GNAQ | |
G-ALPHA-q, GAQ | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.