Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNB3, Mouse, Clone: M1-1-1D5, Abnova™

Catalog No. 89022344 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-344 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-344 Supplier Abnova Corporation Supplier No. H00002784M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant GNB3.

Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors. A single-nucleotide polymorphism (C825T) in this gene is associated with essential hypertension and obesity. This polymorphism is also associated with the occurrence of the splice variant GNB3-s, which appears to have increased activity. GNB3-s is an example of alternative splicing caused by a nucleotide change outside of the splice donor and acceptor sites. Additional splice variants may exist for this gene, but they have not been fully described. [provided by RefSeq

Sequence: MGEMEQLRQEAEQLKKQIADARKACAGVTLAELVSGLEVVGRVQMRTRRTLRGHLAKIYAMHWATDSKLLVSASQDGKLIVWDSYTTNKVHAIPLRSSWVMTCAYAPSGNFVACGGLDNMCSIYNLKSREGNVKVSRELSAHTGYLSCCRFLDDNNIVTSSGDTTCALWDIETGQQKTVFVGHTGDCMSLAVSPDFNLFISGACDASAKLWDVREGTCRQTFTGHESDINAICFFPNGEAICTGSDDASCRLFDLRADQELICFSHESIICSITSVAFSLSGRLLFAGYDDFNCNVWDSMKSERVGILSGHDNRVSCLGVTADGMAVATGSWDSFLKIWN

Specifications

Antigen GNB3
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone M1-1-1D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant GNB3.
Formulation PBS with no preservative; pH 7.4
Gene GNB3
Gene Accession No. BC002454
Gene Symbols GNB3
Host Species Mouse
Immunogen GNB3 (AAH02454, 1 a.a. ∼ 340 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2784
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.