Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

guanine nucleotide binding protein (G protein), gamma 5, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant GNG5.

Supplier:  Abnova Corporation H00002787A01

Catalog No. 89-004-139


Only null left
Add to Cart

Description

Description

Sequence: MSGSSSVAAMKKVVQQLRLEAGLNRVKVSQAAADLKQFCLQNAQHDPLLTGVSSSTNPFRPQKV
Specifications

Specifications

guanine nucleotide binding protein (G protein), gamma 5
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant GNG5.
GNG5
FLJ92393
Mouse
50 μL
Signal Transduction
Primary
Human
Serum
ELISA, Western Blot
Unconjugated
50% glycerol
NM_005274
GNG5
GNG5 (NP_005265, 1 a.a. ∼ 64 a.a) partial recombinant protein with GST tag.
RUO
Yes
2787
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.