Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNGT1, Mouse, Clone: 1F8, Abnova™

Catalog No. 89022350 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-350 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-350 Supplier Abnova Corporation Supplier No. H00002792M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant GNGT1.

Heterotrimeric guanine nucleotide-binding proteins (G proteins) transduce extracellular signals received by transmembrane receptors to effector proteins. Transducin is a guanine nucleotide-binding protein found specifically in rod outer segments, where it mediates activation by rhodopsin of a cyclic GTP-specific (guanosine monophosphate) phosphodiesterase. Transducin is also referred to as GMPase. GNGT1 encodes the gamma subunit of transducin (Hurley et al., 1984 [PubMed 6438626]; Scherer et al., 1996 [PubMed 8661128]).[supplied by OMIM

Sequence: MPVINIEDLTEKDKLKMEVDQLKKEVTLERMLVSKCCEEVRDYVEERSGEDPLVKGIPEDKNPFKELKGGCVIS

Specifications

Antigen GNGT1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant GNGT1.
Formulation PBS with no preservative; pH 7.4
Gene GNGT1
Gene Accession No. BC029367
Gene Alias GNG1
Gene Symbols GNGT1
Host Species Mouse
Immunogen GNGT1 (AAH29367, 1 a.a. ∼ 74 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2792
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.