Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAGK, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127829
Description
N-acetylglucosamine kinase (NAGK; EC 2.7.1.59) converts endogenous N-acetylglucosamine (GlcNAc). a major component of complex carbohydrates. from lysosomal degradation or nutritional sources into GlcNAc 6-phosphate. NAGK belongs to the group of N-acetylhe
PA1-27829 detects GNK from human, mouse, and rat samples.PA1-27829 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human N-Acetylglucosamine kinase variant.
Specifications
GNK | |
Polyclonal | |
NAGK | |
GNK, HSA242910 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
55577 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Western Blot | |
Unconjugated | |
Q9UJ70, Q9QZ08, P81799 | |
NAGK | |
Synthetic peptide corresponding to human N-Acetylglucosamine kinase variant. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction