Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNL3L Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524522
Description
This antibody is predicted to react with mouse based on sequence homology.
The protein encoded by this gene appears to be a nucleolar GTPase that is essential for ribosomal pre-rRNA processing and cell proliferation. Two transcript variants encoding the same protein have been found for this gene.
Specifications
| GNL3L | |
| Polyclonal | |
| GNL3L | |
| guanine nucleotide-binding protein-like 3-like protein, novel GTPase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54552 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Q9NVN8 | |
| GNL3L | |
| KLH conjugated synthetic peptide between 102-130 amino acids from the N-terminal region of human GNL3L | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction