Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNPTG, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89208753 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-208-753 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-208-753 Supplier Abnova Corporation Supplier No. H00084572K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human GNPTG peptide using ARM Technology.

UDP-N-acetylglucosamine:lysosomal-enzyme N-acetylglucosamine-1-phosphotransferase (GlcNAc-phosphotransferase) catalyzes the initial step in the synthesis of the mannose 6-phosphate determinant required for efficient intracellular targeting of newly synthesized lysosomal hydrolases to the lysosome. GlcNAc-phosphotransferase is an alpha-2/beta-2/gamma-2 hexameric complex, the enzyme product of 2 genes, 1 encoded by a gene on chromosome 16 (GNPTG) and the other by a gene on chromosome 12 (GNPTAB; MIM 607840).[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen GNPTG
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description N-acetylglucosamine-1-phosphate transferase, gamma subunit
Formulation In 1X PBS, pH 7.4
Gene GNPTG
Gene Accession No. GNPTG
Gene Alias C16orf27, GNPTAG, LP2537, RJD9
Gene Symbols GNPTG
Host Species Rabbit
Immunogen A synthetic peptide of human GNPTG
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84572
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.