Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOK/Stim1 Mouse, Unlabeled, Clone: 44, BD

Catalog No. BDB610954 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610954 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610954 Supplier BD Biosciences Supplier No. 610954
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The human chromosomal region 11p15 has undergone intense analysis because of its association with various malignancies. In particular, the band 11p15.5 contains genes associated with Wilms tumor, Beckwith-Weidemann syndrome, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. One such gene, GOK (Stim 1), was identified near the 5′ end of the ribonucleotide reductase subunit 1 gene. Examination of the GOK primary amino acid sequence indicates that it is a typical transmembrane protein with an extracellular N-terminal domain and a cytosolic C-terminal domain. The protein is highly hydrophobic with only a short region of hydrophobicity that likely represents the transmembrane region. The C-terminal portion of GOK shares some small regions of homology with myosin (20% identity). This region of GOK consists of α-helices and is thought to adopt a coiled-coil conformation. Although GOK expression has no effect on the growth of certain breast cancer cell lines, it induces death in rhabdomyosarcoma cells. Thus, it is thought to be a recessive tumor suppressor in muscle cells, possibly by functioning as a receptor connected to an apoptotic signaling pathway.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GOK/Stim1
Applications Western Blot
Classification Monoclonal
Clone 44
Concentration 250μg/mL
Conjugate Unconjugated
Description Stim1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human GOK aa. 25-139
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.