Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLGA1, Mouse, Clone: 6G3, Abnova™

Catalog No. 89022353 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-353 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-353 Supplier Abnova Corporation Supplier No. H00002800M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GOLGA1.

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome. [provided by RefSeq]

Sequence: EKPGPEMANMAPSVTNNTDLTDAREINFEYLKHVVLKFMSCRESEAFHLIKAVSVLLNFSQEEENMLKETLEYKMSWFGSKPAPKGSIRPSISNPRIPWS

Specifications

Antigen GOLGA1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6G3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GOLGA1.
Formulation PBS with no preservative; pH 7.4
Gene GOLGA1
Gene Accession No. NM_002077
Gene Alias MGC33154/golgin-97
Gene Symbols GOLGA1
Host Species Mouse
Immunogen GOLGA1 (NP_002068, 668 a.a. ∼ 767 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2800
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.