Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goosecoid (GSC) Rabbit anti-Human, Pig, Rat, Chimpanzee, Rhesus Macaque, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABD15
Description
This Anti-Goosecoid (GSC) Antibody is validated for use in western blot, immunohistochemistry (paraffin) for the detection of Goosecoid (GSC).
Specifications
Goosecoid (GSC) | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Chimpanzee, Human, Pig, Rat, Rhesus Monkey |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
GSC | |
KLH-conjugated linear peptide corresponding to human Goosecoid near the C-terminus. | |
100 μg | |
NP_776248 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction