Learn More
glycoprotein A33 (transmembrane), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human GPA33 protein.
Supplier: Abnova Corporation H00010223B01
Description
The glycoprotein encoded by this gene is a cell surface antigen that is expressed in greater than 95% of human colon cancers. The open reading frame encodes a 319-amino acid polypeptide having a putative secretory signal sequence and 3 potential glycosylation sites. The predicted mature protein has a 213-amino acid extracellular region, a single transmembrane domain, and a 62-amino acid intracellular tail. The sequence of the extracellular region contains 2 domains characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily. [provided by RefSeq
Sequence: MVGKMWPVLWTLCAVRVTVDAISVETPQDVLRASQGKSVTLPCTYHTSTSSREGLIQWDKLLLTHTERVVIWPFSNKNYIHGELYKNRVSISNNAEQSDASITIDQLTMADNGTYECSVSLMSDLEGNTKSRVRLLVLVPPSKPECGIEGETIIGNNIQLTCQSKEGSPTPQYSWKRYNILNQEQPLAQPASGQPVSLKNISTDTSGYYICTSSNEEGTQFCNITVAVRSPSMNVALYVGIAVGVVAALIIIGIIIYCCCCRGKDDNTEDKEDARPNREAYEEPPEQLRELSREREEEDDYRQEEQRSTGRESPDHLDQSpecifications
glycoprotein A33 (transmembrane) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human GPA33 protein. | |
GPA33 | |
A33/MGC129986/MGC129987 | |
Mouse | |
50 μL | |
Membrane Receptors | |
Primary | |
Human | |
Serum |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
No additive | |
NM_005814.1 | |
GPA33 | |
GPA33 (NP_005805.1, 1 a.a. ∼ 319 a.a) full-length human protein. | |
RUO | |
Yes | |
10223 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.