Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPC3 Mouse anti-Human, Mouse, Clone: 9C2, MilliporeSigma™

Catalog No. MABC667MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC667MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC667MI Supplier MilliporeSigma Supplier No. MABC667
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Glypican using this mouse monoclonal antibody, Anti-GPC3 Antibody, clone 9C2 validated for use in western blotting, Immunofluorescence, immunohistochemistry and Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPC3
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 9C2
Conjugate Unconjugated
Formulation Purified mouse monoclonal in PBS containing up to 0.1% sodium azide and 0.5% protein stabilizer.
Gene Symbols GPC3, OCI5
Host Species Mouse
Immunogen Purified recombinant fragment of human GPC3 expressed in E. Coli.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.