Learn More
Description
Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. [provided by RefSeq
Specifications
Specifications
| Antigen | GPC5 |
| Applications | ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 1C9 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant GPC5. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | GPC5 |
| Gene Accession No. | BC039730 |
| Gene Symbols | GPC5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.