Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPCR G2A Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB9860MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9860MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9860MI Supplier MilliporeSigma Supplier No. AB9860
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-GPCR G2A Antibody detects level of GPCR G2A and has been published and validated for use in immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPCR G2A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in 0.1M Glycine / 0.2M Tris buffer solution pH7.8 containing 0.02% sodium azide.
Gene Symbols GPR132; G2A; MGC99642
Host Species Rabbit
Immunogen Highly purified peptide from C-terminus of rat G2A.
Purification Method Affinity Purified
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_013345.2
Target Species Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.