Learn More
Description
Specifications
Specifications
| Antigen | GPCR G2A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 0.1M Glycine / 0.2M Tris buffer solution pH7.8 containing 0.02% sodium azide. |
| Gene Symbols | GPR132; G2A; MGC99642 |
| Host Species | Rabbit |
| Immunogen | Highly purified peptide from C-terminus of rat G2A. |
| Purification Method | Affinity Purified |
| Quantity | 25 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.