Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPIHBP1, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89207455 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-207-455 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-207-455 Supplier Abnova Corporation Supplier No. H00338328K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human GPIHBP1 peptide using ARM Technology.

Dietary fats are packaged by intestine into triglyceride-rich lipoproteins called chylomicrons. The triglycerides in chylomicrons are hydrolyzed by lipoprotein lipase (LPL: MIM 609708) along the luminal surface of capillaries, mainly in heart, skeletal muscle, and adipose tissue. GPIHBP1 is a capillary endothelial cell protein that provides a platform for LPL-mediated processing of chylomicrons (Beigneux et al., 2007 [PubMed 17403372]).[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen GPIHBP1
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1
Formulation In 1X PBS, pH 7.4
Gene GPIHBP1
Gene Accession No. GPIHBP1
Gene Alias GPI-HBP1
Gene Symbols GPIHBP1
Host Species Rabbit
Immunogen A synthetic peptide of human GPIHBP1
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338328
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.