Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR24, Mouse, Clone: 3D7, Abnova™

Catalog No. 89022365 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-365 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-365 Supplier Abnova Corporation Supplier No. H00002847M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GPR24.

The protein encoded by this gene, a member of the G protein-coupled receptor family 1, is an integral plasma membrane protein which binds melanin-concentrating hormone. The encoded protein can inhibit cAMP accumulation and stimulate intracellular calcium flux, and is probably involved in the neuronal regulation of food consumption. Although structurally similar to somatostatin receptors, this protein does not seem to bind somatostatin. [provided by RefSeq

Sequence: VGLGGGSGCQATEEDPLPDCGACAPGQGGRRWRLPQPAWVEGSSARLWEQATGTGWMDLEASLLPTGPNASNTSDGPDNLTSAGSPPRTGSISYINIIMP

Specifications

Antigen GPR24
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GPR24.
Formulation PBS with no preservative; pH 7.4
Gene MCHR1
Gene Accession No. BC001736
Gene Alias GPR24/MCH1R/MGC32129/SLC1
Gene Symbols MCHR1
Host Species Mouse
Immunogen GPR24 (AAH01736, 14 a.a. ∼ 113 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2847
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.