Learn More
GPR Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN473
Description
Detect GPR using this rabbit polyclonal antibody, Anti-GPR Antibody3 validated for use in western blotting, immunohistochemistry and Immunofluorescence.
Specifications
GPR3 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
2-8°C for 12 months |
Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
GPR3; ACCA | |
KLH-conjugated linear peptide corresponding to human GPR3. | |
100 μg | |
Human, Mouse |