Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP160013
Description
GPR6 Polyclonal specifically detects GPR6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GPR6 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
G protein-coupled receptor 6, G-protein coupled receptor 6, Sphingosine 1-phosphate receptor GPR6 | |
Rabbit | |
38 kDa | |
100 μL | |
GPCR | |
2830 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
P46095 | |
GPR6 | |
Synthetic peptides corresponding to GPR6(G protein-coupled receptor 6) The peptide sequence was selected from the N terminal of GPR6. Peptide sequence NASAASLNDSQVVVVAAEGAAAAATAAGGPDTGEWGPPAAAALGAGGGAN. | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Mouse: 92%; Bovine: 85%; Rat: 85%. | |
Human, Mouse, Rat, Bovine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction