Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR81 Rabbit anti-Human, Monkey, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR81.
Supplier: Abnova Corporation PAB16254
Description
G protein-coupled receptors (GPCRs, or GPRs), such as GPR81, contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.[supplied by OMIM
Specifications
GPR81 | |
Polyclonal | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user. | |
GPR81 | |
GPR81 | |
A synthetic peptide (conjugated with KLH) corresponding to human GPR81. | |
RUO | |
27198 | |
Human, Mouse, Rat | |
Liquid |
Immunohistochemistry (PFA fixed) | |
1mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of GPR81. | |
In PBS (0.09% sodium azide) | |
FKSG80/GPR104/TA-GPCR | |
Rabbit | |
50 μg | |
Primary | |
N-terminal extracellular domain of human. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction