Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR81 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP132128
Description
GPR81 Polyclonal specifically detects GPR81 in Human samples. It is validated for Western Blot.Specifications
| GPR81 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| FKSG80, G protein-coupled receptor 104, G protein-coupled receptor 81, GPR104, G-protein coupled receptor 104, G-protein coupled receptor 81, HCA1, HCAR1, LACR1, lactate receptor 1, TA-GPCR, T-cell activation G protein-coupled receptor | |
| Rabbit | |
| 39 kDa | |
| 100 μL | |
| Chemokine Research, GPCR, Immunology | |
| 27198 | |
| Human | |
| IgG |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9BXC0 | |
| HCAR1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human GPR81. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%) and Chimpanzee (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction