Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPS1, Mouse, Clone: 1E8, Abnova™

Catalog No. 89022376 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-376 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-376 Supplier Abnova Corporation Supplier No. H00002873M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GPS1.

This gene is known to suppress G-protein and mitogen-activated signal transduction in mammalian cells. The encoded protein shares significant similarity with Arabidopsis FUS6, which is a regulator of light-mediated signal transduction in plant cells. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: AAAFNTTVAALEDELTQLILEGLISARVDSHSKILYARDVDQRSTTFEKSLLMGKEFQRRAKAMMLRAAVLRNQIHVKSPPREGSQGELTPANSQSRMSTNM

Specifications

Antigen GPS1
Applications ELISA
Classification Monoclonal
Clone 1E8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GPS1.
Formulation PBS with no preservative; pH 7.4
Gene GPS1
Gene Accession No. BC000155
Gene Alias COPS1/CSN1/MGC71287
Gene Symbols GPS1
Host Species Mouse
Immunogen GPS1 (AAH00155, 390 a.a. ∼ 491 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2873
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.