Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

glutamic-pyruvate transaminase (alanine aminotransferase), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017692 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-692 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-692 Supplier Abnova Corporation Supplier No. H00002875D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human GPT protein.

Glutamate-pyruvate transaminase (EC 2.6.1.2), also known as alanine aminotransferase, catalyzes the reversible conversion of L-alanine and alpha-ketoglutarate to L-glutamate and pyruvate. It has 2 distinct molecular and genetic forms: one cytoplasmic (soluble) (GPT1) and one mitochondrial (GPT2; MIM 138210). See ALTQTL1 (MIM 612363) and ALTQTL2 (MIM 612364) for information on quantitative trait loci influencing the plasma level of alanine aminotransferase.[supplied by OMIM

Sequence: MASSTGDRSQAVRHGLRAKVLTLDGMNPRVRRVEYAVRGPIVQRALELEQELRQGVKKPFTEVIRANIGDAQAMGQRPITFLRQVLALCVNPDLLSSPNFPDDAKKRAERILQACGGHSLGAYSVSSGIQLIREDVARYIERRDGGIPADPNNVFLSTGASDAIVTVLKLLVAGEGHTRTGVLIPIPQYPLYSATLAELGAVQVDYYLDEERAWALDVAELHRALGQARDHCRPRALCVINPGNPTGQVQTRECIEAVIRFAFEERLFLLADEVYQDNVYAAGSQFHSFKKVLMEMGPPYAGQQELASFHSTSKGYMGECGFRGGYVEVVNMDAAVQQQMLKLMSVRLCPPVPGQALLDLVVSPPAPTDPSFAQFQAEKQAVLAELAAKAKLTEQVFNEAPGISCNPVQGAMYSFPRVQLPPRAVERAQELGLAPDMFFCLRLLEETGICVVPGSGFGQREGTYHFRMTILPPLEKLRLLLEKLSRFHAKFTLEYS

Specifications

Antigen glutamic-pyruvate transaminase (alanine aminotransferase)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human GPT protein.
Formulation PBS with no preservative; pH 7.4
Gene GPT
Gene Accession No. NM_005309.1
Gene Alias AAT1/ALT1/GPT1
Gene Symbols GPT
Host Species Rabbit
Immunogen GPT (NP_005300.1, 1 a.a. ∼ 496 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2875
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.