Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPVI Rat anti-Human, Clone: 8E9, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABT406
Description
Anti-GPVI Antibody, clone 8E9, Capture Antibody is an antibody against GPVI for use in western blotting and ELISA.
Specifications
GPVI | |
Monoclonal | |
Unconjugated | |
NP_001242946 | |
Rat | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
ELISA, Western Blot | |
8E9 | |
Purified rat monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
GP6 | |
Recombinant protein corresponding to human GPVI. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction