Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-GPx4 Clone: 1B4,

Catalog No. MABS1274MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1274MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1274MI Supplier MilliporeSigma™ Supplier No. MABS1274
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects GPx4 Clone: 1B4 in Mouse samples, and it is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting

Phospholipid hydroperoxide glutathione peroxidase, mitochondrial (EC 1.11.1.12; UniProt O70325; also known as GPx-4, GSHPx-4, mtPHGPx, PHGPx, Phospholipid hydroperoxide glutathione peroxidase mitochondrial, snGPx, Sperm nuclei glutathione peroxidase) is encoded by the Gpx4 gene (Gene ID 625249) in murine species. Gpx4 is a member of the glutathione peroxidase (GPx) family of peroxidases that protect cells against oxidative damage by reducing lipid hydroperoxides to their corresponding alcohols and by reducing free hydrogen peroxide to water. Eight glutathione peroxidase members (GPx1-8) have been identified in human. Inducible Gpx4-knockout in transgenic mice reveals the essential role of Gpx4 in maintaining proper kidney function, and Gpx4 depletion results in massive cell death of renal tubular epithelia in vivo. While the most abundant mammalian GPx, GPx1, that forms tetramers, GPx4 exists in the monomeric form. Three alternatively spliced GPx4 isoforms originate from the same Gpx4 gene, a cytosolic (also called the short form), a mitochondrial (also called the long form), and a nuclear form. The short form is expressed ubiquitously in embryonic and adult tissues in varying degrees, while the expression of the other two forms is restricted to testis where they exert significant functions during male gametogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPx4
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1B4
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O70325
Gene Symbols Gpx4
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to the C-terminal sequence of mouse GPx-4.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001032830.2
Test Specificity Reacts with the Mitochondrial (UniProt O70325-1), Cytoplasmic (UniProt O70325-2), and Nuclear (Q91XR9-1) isoforms.
Target Species Mouse
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.