Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gq/11α Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 06709
Description
This Anti-Gq/11α Antibody, CT is validated for use in immunoprecipitation, western blotting for the detection of Gq/11α.
Specifications
Gq/11α | |
Polyclonal | |
PBS, pH 7.4 | |
Rabbit | |
Protein A chromatography | |
Primary | |
Human, Mouse | |
Purified |
Immunoprecipitation, Western Blot | |
Unconjugated | |
GNA11; GA11; GNA-11 | |
peptide (QLNLKEYNLV) corresponding to amino acids 350-359 of Gqα or G11α | |
50 μL | |
NM_002072 & NM_002067 | |
−20°C for 24 months | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction