Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Gqalpha- and G11alpha-Subunits, C-Terminal, Polyclonal

Catalog No. 37175150UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
37-175-150UL 50 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-175-150UL Supplier MilliporeSigma™ Supplier No. 37175150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Gqα-, and G11α-Subunits, C-Terminal Clone: in samples, and it is validated for ELISA, Immunoprecipitation, Immuno Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Gqα-, and G11α-Subunits, C-Terminal
Applications ELISA, Immunoblot, Immunoblot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In 140mM NaCl, 100mM potassium phosphate, pH 7.0.
Host Species Rabbit
Immunogen a synthetic peptide (QLNLKEYNLV) corresponding to amino acids at the C-terminus of mammalian Gqα and G11α, conjugated to KLH
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the Gqα-and G11α-subunits. Does not cross-react with other G-proteins of the non-pertussis toxin-sensitive family.
Content And Storage ≤ -70°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.