Learn More
Mouse Gr-1/Ly-6G Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Supplier:  R&D Systems FAB1037N100
 View more versions of this product
Catalog No. FAB1037N100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Gr-1/Ly-6G
Monoclonal
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Ly6g
Protein A or G purified from hybridoma culture supernatant
RUO
58530
Mouse
Supernatant
Flow Cytometry
RB6-8C5
Flow Cytometry 5 uL/10^6 cells
C6orf23, G6D, Ly-6C, LY6-D, Ly6G, Ly-6G, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25
Rat
100 Tests
Primary
Detects mouse Gr-1/Ly-6G. Weak cross-reactivity with Ly-6C is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only