Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Granulocytes Mouse anti-Rat, Unlabeled, Clone: RP-1, BD

Catalog No. BDB550000 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550000 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550000 Supplier BD Biosciences Supplier No. 550000
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The RP-1 antibody reacts with rat peritoneal and peripheral blood neutrophils and bone marrow cells. It does not bind rat macrophages, rat eosinophils, or peritoneal neutrophils from mice, rabbits, guinea pigs, or human peripheral blood as determined by antibody adsorption experiments. Flow cytometric analysis of peripheral blood leukocytes indicates that all granulocytes (cells having high side scatter), but not lymphocytes or monocytes (low side-scatter cells), express that antigen recognized by RP-1 mAb. Expression of the RP-1 antigen on rat peritoneal neutrophils is enhanced by stimulation with PMA or ConA. Immunoprecipitation of rat PMA-activated and non-treated neutrophil membranes with the RP-1 mAb produced two bands of approximately 85kDa in MW. However, the RP-1 antigen has not been biochemically characterized.

Flow Cytometry, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen Granulocytes
Applications Flow Cytometry
Classification Monoclonal
Clone RP-1
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen WKA/Hok rat peritoneal neutrophils
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.